Learn More
Medchemexpress LLC Corticosterone | 50-22-6 | 99.8% | 200 MG
SDP
Catalog No. 5000413129
Encompass_Preferred
Corticosterone is an orally active glucocorticoid produced by the adrenal cortex. It is involved in regulating neuronal functions within the limbic system, including the hippocampus, prefrontal cortex, and amygdala.
  • Enhances Rab-mediated AMPAR membrane traffic through SGK-induced phosphorylation of GDI.
  • Has an immunosuppressive effect by interfering with dendritic cell maturation.
Catalog No. 50-004-13129 Supplier Medchemexpress LLC Supplier No. HYB1618200MG
Corticosterone is an orally active glucocorticoid produced by the adrenal cortex. It is involved in regulating neuronal functions within the limbic system, including the hippocampus, prefrontal cortex, and amygdala.
  • Enhances Rab-mediated AMPAR membrane traffic through SGK-induced phosphorylation of GDI.
  • Has an immunosuppressive effect by interfering with dendritic cell maturation.