Learn More
Enzo Life Sciences Corticosterone (50mg). CAS: 50-22-6
SDP
Supplier:  Enzo Life Sciences ALX370007M050
Encompass_Preferred
Alternative name: (11beta)-11,21-Dihydroxypregn-4-ene-3,20-dione, Reichstein's Substance H. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol (50mg/ml) or dioxane (20mg/ml). Long Term Storage: Ambient.
Catalog No. 50-200-8888