Learn More
Medchemexpress LLC Corticosterone-d4 | 2243253-91-8 | 99.2% | 1 MG
SDP
Catalog No. 5000411701
Encompass_Preferred
Corticosterone-d4 is the deuterium labeled Corticosterone. Corticosterone is an adrenocortical steroid that has modest but significant activities as a mineralocorticoid and a glucocorticoid.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-11701 Supplier Medchemexpress LLC Supplier No. HYB1618S11MG
Corticosterone-d4 is the deuterium labeled Corticosterone. Corticosterone is an adrenocortical steroid that has modest but significant activities as a mineralocorticoid and a glucocorticoid.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.