Learn More
Medchemexpress LLC Corticosterone-d8 | 1271728-07-4 | 99.9% | 1 MG
SDP
Catalog No. 5000413060
Encompass_Preferred
Corticosterone-d8 is the deuterium labeled Corticosterone. Corticosterone is an adrenocortical steroid that has modest but significant activities as a mineralocorticoid and a glucocorticoid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13060 Supplier Medchemexpress LLC Supplier No. HYB1618S1MG
Corticosterone-d8 is the deuterium labeled Corticosterone. Corticosterone is an adrenocortical steroid that has modest but significant activities as a mineralocorticoid and a glucocorticoid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.