Learn More
Medchemexpress LLC Corticosterone-d8 | 1271728-07-4 | 99.91% | 5 MG
SDP
Catalog No. 5000413415
Encompass_Preferred
Corticosterone-d8 is the deuterium-labeled Corticosterone. Corticosterone is an adrenocortical steroid that exhibits modest but significant activities as both a mineralocorticoid and a glucocorticoid. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation during drug development, with deuteration specifically recognized for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-13415 Supplier Medchemexpress LLC Supplier No. HYB1618S5MG
Corticosterone-d8 is the deuterium-labeled Corticosterone. Corticosterone is an adrenocortical steroid that exhibits modest but significant activities as both a mineralocorticoid and a glucocorticoid. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation during drug development, with deuteration specifically recognized for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS