Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Corticosterone Double Antibody RIA Kit

Catalog No. p-7162257 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 Tubes
200 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07120102 100 Tubes
MP07120103 200 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP07120102 Supplier MP Biomedical™ Supplier No. 07120102
Add to Cart
Add to Cart

Quantitative Liquid Phase Radioimmunoassay

  • Intended for the measurement of Corticosterone in a sample
  • Specifically desgined for use in laboratory mice and rats
  • Utilizing the combination of a highly specific antiserum for corticosterone and a proprietary blocking function incorporated in the assay system, this RIA is suitable for determination of corticosterone in unextracted serum or plasma without a protein denaturation step.
  • The use of an 125I label eliminates the need for the liquid scintillation counting step required with tritiated assays.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Plasma, Serum
Target Species Mouse, Rat
For Use With (Application) Animal Research
Quantity 100 Tubes
Sample Volume 10 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.