Learn More
Enzo Life Sciences Corticosterone ELISA kit (1x96wells)
SDP
Catalog No. 50658140
Encompass_Preferred
Highly sensitive corticosterone ELISA kit, species independent, used for animal stress research. The Corticosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. The Corticosterone EIA kit has low cross-reactivity with related steroids. The broad dynamic range makes this kit ideal for a wide variety of sample matrices from any species. Sensitivity: 27.0 pg/ml (range 32 - 20,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: DxS IgG Microtiter plate, Assay buffer 15 concentrate, Steroid displacement reagent, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-140 Supplier Enzo Life Sciences Supplier No. ADI900097
Add to Cart
Add to Cart
Highly sensitive corticosterone ELISA kit, species independent, used for animal stress research. The Corticosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. The Corticosterone EIA kit has low cross-reactivity with related steroids. The broad dynamic range makes this kit ideal for a wide variety of sample matrices from any species. Sensitivity: 27.0 pg/ml (range 32 - 20,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: DxS IgG Microtiter plate, Assay buffer 15 concentrate, Steroid displacement reagent, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution