Learn More
Alpco Ltd Mouse/Rat Corticosterone ELISA
SDP
Supplier:  Alpco Ltd 55CORMSE01
Encompass
The Mouse / Rat Corticosterone ELISA is for the quantitative determination of corticosterone in rat and mouse serum or plasma.Includes ready to use standardsRequires 10 ?l of sample per well Incubation time of only 2 hoursSee Rat Control Set 55-DEV99RC for kit controlsFor research use only. Not for use in diagnostic procedures.
Catalog No. 50-751-3631