Learn More
Abcam Corticosterone, endogenous steroid hormone, 50MG
Catalog No. 502706105
Encompass_Preferred
MW 346.5 Da, Purity >98%. Potent endogenous glucocorticoid and mineralocorticoid receptor agonist (EC₅₀ = 20 nM, GR). Induces apoptosis. Modulates carbohydrate, potassium, and sodium metabolism in vivo. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6105 Supplier Abcam Supplier No. AB14359750MG
MW 346.5 Da, Purity >98%. Potent endogenous glucocorticoid and mineralocorticoid receptor agonist (EC₅₀ = 20 nM, GR). Induces apoptosis. Modulates carbohydrate, potassium, and sodium metabolism in vivo. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement