Learn More
Sigma Aldrich Fine Chemicals Biosciences Corticosterone, 50-22-6, MFCD00037715, 100 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 27840100MG
Encompass
Empirical Formula (Hill Notation): C21H30O4, Molecular Weight: 346.46, ≥98.5% (HPLC), Synonym: 11β,21-Dihydroxy-4-pregnene-3,20-dione, 11β,21-Dihydroxyprogesterone, 4-Pregnene-11β,21-diol-3,20-dione, Kendall’s Compound B, Reichstein’s Substance H. Corticosterone was used to study the adrenoceptor signaling in rats, lizards or in vitro. It was also used in immune suppression experiment.
Catalog No. NC0677590