Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Corticotropin-releasing factor (human) | 86784-80-7 | 99.1% | 4757.45 | 10 MG
SDP

Catalog No. 5000426074
Encompass_Preferred

Corticotropin-releasing factor human (Human CRF) stimulates the synthesis and secretion of adrenocorticotropin in the anterior pituitary. It is for research use only and not for sale to patients.

  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Ser-Glu-Glu-Pro-Pro-Ile-Ser-Leu-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Met-Ala-Arg-Ala-Glu-Gln-Leu-Ala-Gln-Gln-Ala-His-Ser-Asn-R-Lys-Leu-Met-Glu-Ile-Ile-NH2
  • Soluble in DMSO at 25 mg/mL
  • Ships at room temperature in continental US
  • Sealed storage, away from moisture and light, under nitrogen. Powder stable for 2 years at -80°C, 1 year at -20°C. In solvent, stable for 6 months at -80°C, 1 month at -20°C.

Catalog No. 50-004-26074 Supplier Medchemexpress LLC Supplier No. HYP008610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Corticotropin-releasing factor human (Human CRF) stimulates the synthesis and secretion of adrenocorticotropin in the anterior pituitary. It is for research use only and not for sale to patients.

  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Ser-Glu-Glu-Pro-Pro-Ile-Ser-Leu-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Met-Ala-Arg-Ala-Glu-Gln-Leu-Ala-Gln-Gln-Ala-His-Ser-Asn-R-Lys-Leu-Met-Glu-Ile-Ile-NH2
  • Soluble in DMSO at 25 mg/mL
  • Ships at room temperature in continental US
  • Sealed storage, away from moisture and light, under nitrogen. Powder stable for 2 years at -80°C, 1 year at -20°C. In solvent, stable for 6 months at -80°C, 1 month at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.