Learn More
Carbosynth LLC. Mouse monoclonal Cortisol antibody
SDP
Supplier:  Carbosynth LLC. 10C30D1MG
Encompass_Preferred
Cortisol (hydrocortisone) is a steroid hormone, or glucocorticoid, produced by the adrenal gland. It is released in response to stress and a low level of blood glucocorticoids. Its primary functions are to increase blood sugar through gluconeogenesis; suppress the immune system; and aid in fat, protein and carbohydrate metabolism. It also decreases bone formation. During pregnancy, increased production of cortisol between weeks 30-32 initiates production of fetal lung surfactanct to promote maturation of the lungs. Various synthetic forms of cortisol are used to treat a variety of different diseases.
Catalog No. 50-125-2536