Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cortisol Competitive ELISA Kit

Catalog No. p-200006751
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAHCOR 96 Tests
EIAHCORX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIAHCOR Supplier Invitrogen™ Supplier No. EIAHCOR
Only null left
Add to Cart
Edge
Add to Cart

ELISA

The Human Cortisol ELISA quantitates Cortisol in human serum,plasma,urine,saliva,dried fecal extracts or or cell culture medium.

Principle of the method

The Cortisol Competitive ELISA research-use-only kit is designed to quantitatively measure cortisol present in dried fecal extracts,saliva,urine,serum,plasma,and tissue culture media samples. Total cortisol is measured in extracted samples and in serum and plasma,and free cortisol in saliva and urine. A cortisol standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture mouse antibodies. A cortisol-peroxidase conjugate is added to the wells. The binding reaction is initiated by the addition of a monoclonal antibody to cortisol. After a 1-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound cortisol-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Cortisol is the most potent glucocorticoid produced by the human adrenal. It is synthesized from cholesterol and its production is stimulated by pituitary adrenocorticotropic hormone (ACTH) which is regulated by corticotropin releasing factor (CRF). ACTH and CRF secretions are inhibited by high cortisol levels in a negative feedback loop. In plasma a majority of cortisol is bound with high affinity to corticosteroid binding globulin (CBG or transcotin). Cortisol acts through specific intracellular receptors and affects numerous physiologic systems including immune function, glucose counter regulation, vascular tone, and bone metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 100 to 3200 pg/mL
Assay Sensitivity 17.3 pg/mL
Description Cortisol Competitive ELISA Kit
Assay Multiplex assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Product Type ELISA
Protein Family Steroid hormone
Sample Type Fecal Extract, Plasma, Saliva, Serum, Supernatant, Urine
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 8.1%
Intraassay CV 8.8
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Inflammation
Sample Volume Fecal Extract 50 μL, Plasma 1 μL, Saliva 12.5 μL, Serum 1 μL, Supernatant 50 μL, Urine 6.25 μL
Storage Requirements -20°C
Target Cortisol
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.