Learn More
Alpco Ltd Cortisol ELISA
SDP
Supplier:  Alpco Ltd 11CRLHUE01
Encompass_Preferred
ELISA for the quantitative determination of Cortisol in human serum. Includes ready to use standards, controls, and conjugateSample pretreatment not requiredShort incubation time of less than 2 hours.
Catalog No. NC2299698