Learn More
Enzo Life Sciences Cortisol ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900071
Encompass_Preferred
Widely used sensitive cortisol ELISA kit with a large variety of sample types and highly reproducible results for stress, autoimmune disease and cancer research. The Cortisol EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit is highly cited in peer-reviewed publications. The ready-to-use liquid color-coded reagents provided are safe, non-radioactive, and reduce error in the lab. The broad dynamic range lets you accurately measure Cortisol levels in a variety of sample matrices. Sensitivity: 56.72 pg/ml (range 156 - 10,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent
Catalog No. 50-658-129