Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedical™ Cortisol ELISA Kit
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 x 96 Wells
2 x 96 Wells
Description
The Cortisol ELISA Kit is intended for the measurement of total cortisol concentration in human serum or plasma by a microplate enzyme immunoassay.
Specifications
Specifications
| Product Type | Kit and Assay |
| Sample Type | Serum |
| Target Species | Human |
| For Use With (Application) | Endocrinology |
| Quantity | 1 x 96 Wells |
| Storage Requirements | 2°C to 8°C |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction