Learn More
Cayman Chemical Cortisol ELISA Kit, 480 strip wells
SDP
Supplier:  Cayman Chemical 500360480STRIPWELLS
Encompass_Preferred
Cayman’s Cortisol ELISA is a competitive assay that provides accurate measurements of cortisol within the range of 6.6-4,000 pg/ml
Catalog No. NC1468473
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.