Learn More
Abcam Cortisol ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1549961X96TESTS
Encompass_Preferred
Cortisol ELISA Kit ab154996 is used to quantify Cortisol with a sensitivity of 2.9 pg/ml. Competitive ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2944