Learn More
Drg International Inc DRG Cortisol ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA1887R
Encompass_Preferred
An enzyme immunoassay for the quantitative determination of cortisol in serum and plasma. This product is for Research Use Only. The DRG Cortisol ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a monoclonal antibody directed towards an antigenic site on the Cortisol molecule.  Endogenous Cortisol of a sample competes with a Cortisol-horseradish peroxidase conjugate for binding to the coated antibody.
Catalog No. 50-187-1244