Learn More
Biomatik Corporation Cortisol Monoclonal Antibody
Supplier:  Biomatik Corporation CAC11881
Encompass_Preferred
Cortisol Monoclonal Antibody is a Monoclonal Antibody to Cortisol. The Cortisol Monoclonal Antibody has been validated for the following applications: ELISA. Host: Mouse. Immunogen: Cortisol-Bsa Conjugate. Applications: ELISA. Target Synonyms: Cortisol. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9234