Learn More
Medchemexpress LLC Cortisol sulfate sodium | 1852-36-4 | 1 ML
SDP
Catalog No. 5000417424
Encompass_Preferred
Cortisol sulfate sodium, also known as Cortisol 21-sulfate sodium, is a metabolite of Cortisol. It functions as a specific ligand for intracellular transcortin and has been observed to affect behavior through direct action on the central nervous system.
  • Metabolite of Cortisol
  • Specific ligand for intracellular transcortin
  • Affects behavior via central nervous system action
Catalog No. 50-004-17424 Supplier Medchemexpress LLC Supplier No. HYN8460A10MM/1ML
Cortisol sulfate sodium, also known as Cortisol 21-sulfate sodium, is a metabolite of Cortisol. It functions as a specific ligand for intracellular transcortin and has been observed to affect behavior through direct action on the central nervous system.
  • Metabolite of Cortisol
  • Specific ligand for intracellular transcortin
  • Affects behavior via central nervous system action