Learn More
Cayman Chemical Cortisone Acetate, Purity: ≥98%, CAS: 50-04-4, 25 g
SDP
Catalog No. 502995102
Encompass_Preferred
A synthetic glucocorticoid with anti-inflammatory properties; decreases the size of BCG vaccine-induced dermal lesions and tuberculin reactions in rabbits (2 mg/kg on alternate days over the course of 46 days); slows tissue regeneration in a rabbit model of wound healing (2.5 mg/kg per day, s.c., over 4 weeks); decreases the number of dexamethasone binding sites on isolated human lymphocytes by 30%
Catalog No. 50-299-5102 Supplier Cayman Chemical Supplier No. 2379825 G
A synthetic glucocorticoid with anti-inflammatory properties; decreases the size of BCG vaccine-induced dermal lesions and tuberculin reactions in rabbits (2 mg/kg on alternate days over the course of 46 days); slows tissue regeneration in a rabbit model of wound healing (2.5 mg/kg per day, s.c., over 4 weeks); decreases the number of dexamethasone binding sites on isolated human lymphocytes by 30%