Learn More
Medchemexpress LLC Cortisone 21-acetate (Standard) | 50-04-4 | 99.4% | 402.48 | 100 MG
SDP
Catalog No. 5000386637
Encompass_Preferred
Cortisone acetate (Standard) is the analytical standard of Cortisone acetate, intended for research and analytical applications. It is an oxidized metabolite of Cortisol (a Glucocorticoid) and functions as an immunosuppressant and anti-inflammatory agent. At high concentrations, Cortisone acetate can partially interfere with the binding of Glucocorticoid to the Glucocorticoid-receptor.
  • Grade of analytical standard, supplied as a reference standard assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS.
  • Acts as an immunosuppressant.
  • Acts as an anti-inflammatory agent.
  • Purity of 99.39%.
Catalog No. 50-003-86637 Supplier Medchemexpress LLC Supplier No. HY17461AR100MG
Cortisone acetate (Standard) is the analytical standard of Cortisone acetate, intended for research and analytical applications. It is an oxidized metabolite of Cortisol (a Glucocorticoid) and functions as an immunosuppressant and anti-inflammatory agent. At high concentrations, Cortisone acetate can partially interfere with the binding of Glucocorticoid to the Glucocorticoid-receptor.
  • Grade of analytical standard, supplied as a reference standard assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS.
  • Acts as an immunosuppressant.
  • Acts as an anti-inflammatory agent.
  • Purity of 99.39%.