Learn More
Selleck Chemical LLC Cortisone
SDP
Supplier:  Selleck Chemical LLC S58691G
Encompass_Preferred
Cortisone is a pregnane steroid hormone and one of main substance released by the body as a reaction to stress It suppresses the immune system thus reducing inflammation and attendant pain and swelling at the site of the injury
Catalog No. 50-000-03835