Learn More
Medchemexpress LLC Cortisone | 53-06-5 | 99.3% | 360.44 | 1 ML
SDP
Catalog No. 5000386431
Encompass_Preferred
Cortisone (17-Hydroxy-11-dehydrocorticosterone) is an oxidized metabolite of Cortisol, a Glucocorticoid. It functions as an immunosuppressant and anti-inflammatory agent. Cortisone can partially intervene in the binding of Glucocorticoid to the Glucocorticoid-receptor at high concentrations, and has been shown to attenuate apoptosis induced by Cortisol in peripheral-blood mononuclear cells (PBMCs) and decrease BCG lesions and tuberculin reactions in rabbits.
  • Acts as an immunosuppressant
  • Acts as an anti-inflammatory agent
  • Attenuates apoptosis
  • Decreases BCG lesions and tuberculin reactions
  • Targets Glucocorticoid Receptor
  • Is an endogenous metabolite
Catalog No. 50-003-86431 Supplier Medchemexpress LLC Supplier No. HY1746110MM/1ML
Cortisone (17-Hydroxy-11-dehydrocorticosterone) is an oxidized metabolite of Cortisol, a Glucocorticoid. It functions as an immunosuppressant and anti-inflammatory agent. Cortisone can partially intervene in the binding of Glucocorticoid to the Glucocorticoid-receptor at high concentrations, and has been shown to attenuate apoptosis induced by Cortisol in peripheral-blood mononuclear cells (PBMCs) and decrease BCG lesions and tuberculin reactions in rabbits.
  • Acts as an immunosuppressant
  • Acts as an anti-inflammatory agent
  • Attenuates apoptosis
  • Decreases BCG lesions and tuberculin reactions
  • Targets Glucocorticoid Receptor
  • Is an endogenous metabolite