Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortisone

Catalog No. aaj6418203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6418203 1 g
AAJ6418206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6418203 Supplier Thermo Scientific Chemicals Supplier No. J6418203
Only null left
Add to Cart

CAS: 53-06-5 | C21H28O5 | 360.45 g/mol

A steroid and immunosuppressant

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-06-5
Molecular Formula C21H28O5
Molecular Weight (g/mol) 360.45
MDL Number MFCD00003610
InChI Key MFYSYFVPBJMHGN-ZPOLXVRWSA-N
Synonym cortisone, andreson, cortisal, cortisate, cortistal, cortivite, cortogen, cortone, kendall's compound e, adrenalex
PubChem CID 222786
ChEBI CHEBI:16962
IUPAC Name (1R,3aS,3bS,9aR,9bS,11aS)-1-hydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-7,10-dione
SMILES C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

Melting Point 223°C to 228°C (decomposition)
Color White
Quantity 1 g
Beilstein 1356062
Merck Index 14,2539
Formula Weight 360.44
Physical Form Powder
Chemical Name or Material Cortisone
Hazard Category H361d
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 200-162-4
RTECSNumber GM9020000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.