Learn More
Apexbio Technology LLC Cortisone acetate 50-04-4 1g
SDP
Catalog No. 5000089785
Encompass_Preferred
Cortisone acetate (50-04-4) is a glucocorticoid receptor agonist targeting the glucocorticoid receptor It is designed to modulate glucocorticoid signaling thereby regulating gene transcription and downstream immune and inflammatory responses Cortisone acetate exerts its biological activity primarily through enzymatic conversion to cortisol which subsequently activates glucocorticoid receptors to influence cellular stress and inflammatory pathways Based on these pharmacological properties cortisone acetate holds research potential in the investigation of glucocorticoid signaling pathways receptor-mediated transcriptional regulation cellular stress responses and inflammatory condition models
Catalog No. 50-000-89785 Supplier Apexbio Technology LLC Supplier No. B19191000
Cortisone acetate (50-04-4) is a glucocorticoid receptor agonist targeting the glucocorticoid receptor It is designed to modulate glucocorticoid signaling thereby regulating gene transcription and downstream immune and inflammatory responses Cortisone acetate exerts its biological activity primarily through enzymatic conversion to cortisol which subsequently activates glucocorticoid receptors to influence cellular stress and inflammatory pathways Based on these pharmacological properties cortisone acetate holds research potential in the investigation of glucocorticoid signaling pathways receptor-mediated transcriptional regulation cellular stress responses and inflammatory condition models