Learn More
Medchemexpress LLC Cortisone acetate | 50-04-4 | 98.9% | 402.48 | 10 G
SDP
Catalog No. 5000386262
Encompass_Preferred
Cortisone acetate (Cortisone 21-acetate) is an oxidized metabolite of Cortisol (a Glucocorticoid). It functions as an immunosuppressant and anti-inflammatory agent. Cortisone acetate can partially intervene in the binding of Glucocorticoid to the Glucocorticoid-receptor at high concentrations.
  • Acts as an immunosuppressant and anti-inflammatory agent
  • Can partially intervene in the binding of Glucocorticoid to Glucocorticoid-receptor
Catalog No. 50-003-86262 Supplier Medchemexpress LLC Supplier No. HY17461A10G
Cortisone acetate (Cortisone 21-acetate) is an oxidized metabolite of Cortisol (a Glucocorticoid). It functions as an immunosuppressant and anti-inflammatory agent. Cortisone acetate can partially intervene in the binding of Glucocorticoid to the Glucocorticoid-receptor at high concentrations.
  • Acts as an immunosuppressant and anti-inflammatory agent
  • Can partially intervene in the binding of Glucocorticoid to Glucocorticoid-receptor