Learn More
Medchemexpress LLC Cortisone acetate | 50-04-4 | 99.9% | 402.48 | 25 G
SDP
Catalog No. 5000385311
Encompass_Preferred
Cortisone acetate, also known as Cortisone 21-acetate, is an oxidized metabolite of Cortisol, which is a Glucocorticoid. It functions as an immunosuppressant and anti-inflammatory agent. At high concentrations, cortisone acetate can partially interfere with the binding of Glucocorticoid to the Glucocorticoid-receptor.
  • Functions as an immunosuppressant.
  • Acts as an anti-inflammatory agent.
  • Is an oxidized metabolite of cortisol.
  • Can partially intervene in binding of glucocorticoid to glucocorticoid-receptor at high concentrations.
Catalog No. 50-003-85311 Supplier Medchemexpress LLC Supplier No. 25GHY17461A
Cortisone acetate, also known as Cortisone 21-acetate, is an oxidized metabolite of Cortisol, which is a Glucocorticoid. It functions as an immunosuppressant and anti-inflammatory agent. At high concentrations, cortisone acetate can partially interfere with the binding of Glucocorticoid to the Glucocorticoid-receptor.
  • Functions as an immunosuppressant.
  • Acts as an anti-inflammatory agent.
  • Is an oxidized metabolite of cortisol.
  • Can partially intervene in binding of glucocorticoid to glucocorticoid-receptor at high concentrations.