Learn More
Medchemexpress LLC Cortisone acetate | 50-04-4 | 99.9% | 402.48 | 5 G
SDP
Catalog No. 5000386636
Encompass_Preferred
Cortisone acetate is an oxidized metabolite of Cortisol. It acts as an immunosuppressant and anti-inflammatory agent. At high concentrations, it can partially intervene in the binding of Glucocorticoid to the Glucocorticoid-receptor.
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Steroids
  • Initial source: Endogenous metabolite
Catalog No. 50-003-86636 Supplier Medchemexpress LLC Supplier No. HY17461A5G
Cortisone acetate is an oxidized metabolite of Cortisol. It acts as an immunosuppressant and anti-inflammatory agent. At high concentrations, it can partially intervene in the binding of Glucocorticoid to the Glucocorticoid-receptor.
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Steroids
  • Initial source: Endogenous metabolite