Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cortisone acetate | 50-04-4 | MFCD00003609 | >99.0% | 402.48 | C23H30O6 | 10 MG
SDP

Catalog No. 5000299101
Encompass_Preferred

Cortisone acetate analytical standard supplied for research and analytical applications. It is used as a reference material in chromatographic and mass spectrometric assays, supporting qualitative, quantitative, and methodological studies.

  • Analytical standard for HPLC, GC, and MS
  • Intended for qualitative and quantitative method development
  • Purity ≥99.0%
  • Molecular weight 402.48 g/mol
  • CAS number 50-04-4
  • Provided as a 10 mg pack suitable for laboratory use
  • Exhibits immunosuppressant and anti-inflammatory activity

Catalog No. 50-002-99101 Supplier Medchemexpress LLC Supplier No. HY17461AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cortisone acetate analytical standard supplied for research and analytical applications. It is used as a reference material in chromatographic and mass spectrometric assays, supporting qualitative, quantitative, and methodological studies.

  • Analytical standard for HPLC, GC, and MS
  • Intended for qualitative and quantitative method development
  • Purity ≥99.0%
  • Molecular weight 402.48 g/mol
  • CAS number 50-04-4
  • Provided as a 10 mg pack suitable for laboratory use
  • Exhibits immunosuppressant and anti-inflammatory activity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.