Learn More
Medchemexpress LLC Cortisone acetate (Standard) | 50-04-4 | 99.4% | 402.48 | 50 MG
SDP
Catalog No. 5000386626
Encompass_Preferred
Cortisone acetate (Standard) serves as an analytical standard for research and analytical applications. This compound, an oxidized metabolite of Cortisol, functions as an immunosuppressant and anti-inflammatory agent. It is commonly utilized in qualitative, quantitative, and methodological research experiments across various techniques.
  • Analytical standard for research
  • Reference standard for assays
  • Immunosuppressant properties
  • Anti-inflammatory agent
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-003-86626 Supplier Medchemexpress LLC Supplier No. HY17461AR50MG
Cortisone acetate (Standard) serves as an analytical standard for research and analytical applications. This compound, an oxidized metabolite of Cortisol, functions as an immunosuppressant and anti-inflammatory agent. It is commonly utilized in qualitative, quantitative, and methodological research experiments across various techniques.
  • Analytical standard for research
  • Reference standard for assays
  • Immunosuppressant properties
  • Anti-inflammatory agent
  • Used in HPLC, GC, and MS experiments