Learn More
Medchemexpress LLC Cortisone-d8 | 1257650-98-8 | 98.2% | 368.49 | 1 MG
SDP
Catalog No. 5000386696
Encompass_Preferred
Cortisone-d8 is the deuterium labeled Cortisone, an oxidized metabolite of Cortisol (a Glucocorticoid). It acts as an immunosuppressant and anti-inflammatory agent, and can partially intervene in binding of Glucocorticoid to Glucocorticoid-receptor at high concentrations. This compound is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-86696 Supplier Medchemexpress LLC Supplier No. HY17461S11MG
Cortisone-d8 is the deuterium labeled Cortisone, an oxidized metabolite of Cortisol (a Glucocorticoid). It acts as an immunosuppressant and anti-inflammatory agent, and can partially intervene in binding of Glucocorticoid to Glucocorticoid-receptor at high concentrations. This compound is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS