Learn More
Medchemexpress LLC Cortisone (Standard) | 53-06-5 | 98.5% | 360.44 | 100 MG
Supplier: Medchemexpress LLC HY17461R100MG
Cortisone (Standard) is the analytical standard of Cortisone, intended for research and analytical applications. It is an oxidized metabolite of Cortisol (a Glucocorticoid), and acts as an immunosuppressant and anti-inflammatory agent. This product is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS, and can partially intervene in the binding of Glucocorticoid to Glucocorticoid-receptor at high concentrations.
- Analytical standard for Cortisone.
- Intended for research and analytical applications.
- Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
- Acts as an immunosuppressant and anti-inflammatory agent.