Learn More
Medchemexpress LLC Cortisone (Standard) | 53-06-5 | 98.5% | 360.44 | 25 MG
SDP
Catalog No. 5000383981
Encompass_Preferred
Cortisone (Standard) is the analytical standard of Cortisone, intended for research and analytical applications. Cortisone (17-Hydroxy-11-dehydrocorticosterone), an oxidized metabolite of Cortisol (a Glucocorticoid), acts as an immunosuppressant and anti-inflammatory agent. It can partially intervene in the binding of Glucocorticoid to Glucocorticoid-receptor at high concentrations. This compound is a grade of analytical standard, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Immunosuppressant and anti-inflammatory agent
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-83981 Supplier Medchemexpress LLC Supplier No. 25MGHY17461R
Cortisone (Standard) is the analytical standard of Cortisone, intended for research and analytical applications. Cortisone (17-Hydroxy-11-dehydrocorticosterone), an oxidized metabolite of Cortisol (a Glucocorticoid), acts as an immunosuppressant and anti-inflammatory agent. It can partially intervene in the binding of Glucocorticoid to Glucocorticoid-receptor at high concentrations. This compound is a grade of analytical standard, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Immunosuppressant and anti-inflammatory agent
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS