Learn More
CPC Scientific H-Pro-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Ser-Ser-Cys-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CORT002A
Encompass
Neuropeptide similar to somatostatin-14 that is mainly expressed in the cortex and shares many functional properties with somatostatin-14 but differs in its sleep modulatory properties.SEQUENCE: H-Pro-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Ser-Ser-Cys-Lys-OH (trifluoroacetate salt)(Cys2 and 13 bridge)ONE-LETTER SEQUENCE: PCKNFFWKTFSSCK (Cys2 and 13 bridge)MOLECULAR FORMULA: C81H113N19O19S2MOLECULAR WEIGHT: 1721.1STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [186901-48-4]SYNONYMS: RESEARCH AREA: NeuropeptidesREFERENCES:L. de Leccea et al., Nature, 381, 242 (1996)S.Fukusumi et al., Biochem. Biophys. Res. Commun., 232, 157 (1997)SKU(s): CORT-002A, CORT-002B
Catalog No. 50-210-9139