Learn More
Medchemexpress LLC HY-P1932A 1mg Medchemexpress, Cortistatin-14 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1932A1MG
Encompass_Preferred
Medchemexpress, HY-P1932A 1mg Cortistatin-14 (TFA) CAS: Cortistatin-14 (TFA), a neuropeptide have structural similarity to somatostatin-14, binds and exerts its function via the somatostatin receptors (sst1-sst5). Cortistatin-14 (TFA) shows anticonvulsive, neuroprotective effect and remarkable anti-inflammatory properties[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0546