Learn More
Medchemexpress LLC Cortistatin 29 (reduced) | 98.1% | 3539.77 | 1 MG
SDP
Catalog No. 5000400169
Encompass_Preferred
Cortistatin 29 (reduced) is a neuropeptide that plays a role in alleviating neuropathic pain. It demonstrates high-affinity binding to all somatostatin (SS) receptor subtypes and also exhibits anti-fibrotic effects.
  • Alleviates neuropathic pain
  • Binds to all somatostatin receptor subtypes
  • Exhibits anti-fibrotic effects
  • High affinity binding to SSTR1 (2.8 nM), SSTR2 (7.1 nM), SSTR3 (0.2 nM), SSTR4 (3.0 nM), and SSTR5 (13.7 nM)
Catalog No. 50-004-00169 Supplier Medchemexpress LLC Supplier No. HYP36181MG
Cortistatin 29 (reduced) is a neuropeptide that plays a role in alleviating neuropathic pain. It demonstrates high-affinity binding to all somatostatin (SS) receptor subtypes and also exhibits anti-fibrotic effects.
  • Alleviates neuropathic pain
  • Binds to all somatostatin receptor subtypes
  • Exhibits anti-fibrotic effects
  • High affinity binding to SSTR1 (2.8 nM), SSTR2 (7.1 nM), SSTR3 (0.2 nM), SSTR4 (3.0 nM), and SSTR5 (13.7 nM)