Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cortistatin-29 (reduced) | 98.1% | 3539.77 | 10 MG
SDP

Catalog No. 5000400001
Encompass_Preferred

Cortistatin-29 (reduced) is a peptide with a molecular weight of 3539.77 and a purity of 98.1% (HPLC). It appears as a white to off-white solid and is identified for use in laboratory chemical applications and the manufacture of substances. This product is for research use only.

  • High purity of 98.1% (HPLC)
  • Provided as a white to off-white solid
  • Suitable for laboratory chemical applications
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month
  • Store sealed, away from moisture and light

Catalog No. 50-004-00001 Supplier Medchemexpress LLC Supplier No. HYP361810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cortistatin-29 (reduced) is a peptide with a molecular weight of 3539.77 and a purity of 98.1% (HPLC). It appears as a white to off-white solid and is identified for use in laboratory chemical applications and the manufacture of substances. This product is for research use only.

  • High purity of 98.1% (HPLC)
  • Provided as a white to off-white solid
  • Suitable for laboratory chemical applications
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month
  • Store sealed, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.