Learn More
Medchemexpress LLC Cortodoxone-d2 | 1271728-08-5 | 99.0% | C21H28D2O4 | 500 UG
SDP
Catalog No. 5000435028
Encompass_Preferred
Cortodoxone-d2 is the deuterium labeled form of Cortodoxone, a glucocorticoid steroid hormone that can be oxygenated to cortisol. Deuteration can affect pharmacokinetic and metabolic profiles, making this compound suitable as a tracer or internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-35028 Supplier Medchemexpress LLC Supplier No. HY77839S1500UG
Add to Cart
Add to Cart
Cortodoxone-d2 is the deuterium labeled form of Cortodoxone, a glucocorticoid steroid hormone that can be oxygenated to cortisol. Deuteration can affect pharmacokinetic and metabolic profiles, making this compound suitable as a tracer or internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS