Learn More
Medchemexpress LLC Cortodoxone-d7 (11-deoxycortisol-d7) | 152-58-9 | MFCD00003662 | 353.5 g/mol | C21H23D7O4 | 1 MG
SDP
Catalog No. 5000284243
Encompass_Preferred
Cortodoxone-d7 is the deuterium-labeled (d7) isotopologue of cortodoxone (11-deoxycortisol) supplied as a purified analytical standard for research use. It is intended for use as a stable isotope internal standard in quantitative mass spectrometry and isotope-dilution assays, enabling accurate analyte quantitation and compensation for matrix effects. Molecular formula C21H23D7O4; molecular weight 353.5 g/mol.
  • Deuterium-labeled analog of 11-deoxycortisol for use as an internal standard.
  • Suitable for LC-MS/MS and isotope-dilution quantitation workflows.
  • Matches chromatographic behavior and ionization of the unlabeled analyte.
  • Molecular formula C21H23D7O4; molecular weight 353.5 g/mol.
  • Provided as a purified reference standard for analytical and research use.
Catalog No. 50-002-84243 Supplier Medchemexpress LLC Supplier No. HYW6539851MG
Cortodoxone-d7 is the deuterium-labeled (d7) isotopologue of cortodoxone (11-deoxycortisol) supplied as a purified analytical standard for research use. It is intended for use as a stable isotope internal standard in quantitative mass spectrometry and isotope-dilution assays, enabling accurate analyte quantitation and compensation for matrix effects. Molecular formula C21H23D7O4; molecular weight 353.5 g/mol.
  • Deuterium-labeled analog of 11-deoxycortisol for use as an internal standard.
  • Suitable for LC-MS/MS and isotope-dilution quantitation workflows.
  • Matches chromatographic behavior and ionization of the unlabeled analyte.
  • Molecular formula C21H23D7O4; molecular weight 353.5 g/mol.
  • Provided as a purified reference standard for analytical and research use.