Learn More
Cayman Chemical Cortol 10mg
SDP
Catalog No. 502949501
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid has been detected in human hair samples tested for verification of drug abstinence intended for research and forensic applications
Catalog No. 50-294-9501 Supplier Cayman Chemical Supplier No. 33412 10MG
An analytical reference standard categorized as a synthetic cannabinoid has been detected in human hair samples tested for verification of drug abstinence intended for research and forensic applications