Learn More
Cayman Chemical Cortol 50mg
SDP
Catalog No. 502949262
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids has been found in Cannabis intended for research and forensic applications
Catalog No. 50-294-9262 Supplier Cayman Chemical Supplier No. 33412 50MG
An analytical reference standard that is structurally similar to known phytocannabinoids has been found in Cannabis intended for research and forensic applications