Learn More
Cayman Chemical β-Cortolone, Purity: ≥95%, CAS: 667-66-3, 10 mg
SDP
Catalog No. 502912775
Encompass_Preferred
A metabolite of cortisol; urinary levels are decreased in postmenopausal women with endometrial adenocarcinoma compared to postmenopausal women without endometrial adenocarcinoma
Catalog No. 50-291-2775 Supplier Cayman Chemical Supplier No. 3622410 MG
A metabolite of cortisol; urinary levels are decreased in postmenopausal women with endometrial adenocarcinoma compared to postmenopausal women without endometrial adenocarcinoma