Learn More
TARGETMOL CHEMICALS INC CORYLIFOL A 10MG
SDP
Catalog No. 502579837
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Corylifol A (Corylinin) displays cytotoxic activity against HepG2 and Hep3B hepatocellular carcinoma cell lines with IC50 values of 4.6 and 13.5 ug/ml respectively. 2. Corylifol A and Biochanin A can be the potential uncouplers of neuronal nitric oxide synthase-postsynaptic density protein-95. 3. Corylifol A and bavachin are strong inhibitors of UDP-glucuronosyltransferase 1A1 (UGT1A1) with the inhibition kinetic parameters (Ki) values lower than 1 uM.4. Corylifol A and bakuchiol are naturally occurring potent inhibitors of hCE2 with low Ki values ranging from 0.62uM to 3.89 uM. 5. Corylifol A shows an inhibitory effect on IL-6-induced STAT3 promoter activity in Hep3B cells with IC50 values of 0.81 ± 0.15 uu it also inhibits STAT3 phosphorylation induced by IL-6 in Hep3B cells suggests that corylifol A has antiinflammatory activity. purity: 99%
Catalog No. 50-257-9837 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S014510MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Corylifol A (Corylinin) displays cytotoxic activity against HepG2 and Hep3B hepatocellular carcinoma cell lines with IC50 values of 4.6 and 13.5 ug/ml respectively. 2. Corylifol A and Biochanin A can be the potential uncouplers of neuronal nitric oxide synthase-postsynaptic density protein-95. 3. Corylifol A and bavachin are strong inhibitors of UDP-glucuronosyltransferase 1A1 (UGT1A1) with the inhibition kinetic parameters (Ki) values lower than 1 uM.4. Corylifol A and bakuchiol are naturally occurring potent inhibitors of hCE2 with low Ki values ranging from 0.62uM to 3.89 uM. 5. Corylifol A shows an inhibitory effect on IL-6-induced STAT3 promoter activity in Hep3B cells with IC50 values of 0.81 ± 0.15 uu it also inhibits STAT3 phosphorylation induced by IL-6 in Hep3B cells suggests that corylifol A has antiinflammatory activity. purity: 99%