Learn More
Medchemexpress LLC HY-N0897 5mg Medchemexpress, Corylifol A CAS:775351-88-7 Purity:>98%
SDP
Catalog No. 501933782
Encompass_Preferred
Medchemexpress, HY-N0897 5mg Corylifol A CAS:775351-88-7 Corylifol A inhibits IL-6-induced STAT3 activation and phosphorylation, with an IC 50 of 0.81 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3782 Supplier Medchemexpress LLC Supplier No. HYN08975MG
Medchemexpress, HY-N0897 5mg Corylifol A CAS:775351-88-7 Corylifol A inhibits IL-6-induced STAT3 activation and phosphorylation, with an IC 50 of 0.81 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.