Learn More
ATCC Corynebacterium accolens
Supplier:  ATCC 49725
Encompass
Corynebacterium accolens strain CNCTC Th 1/57 was isolated from the cervix of a human female in Havlickuv Brod, Czech Republic. This bacterial type strain is whole-genome sequenced and can be propagated aerobically at 37°C.
Catalog No. 50-237-9915