Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 25 (d25F) 10mg
Catalog No. 502381638
Encompass
Purified pneumococcal polysaccharide type 25F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 25F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-1638 Supplier ATCC Supplier No. 531X
Purified pneumococcal polysaccharide type 25F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 25F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.