Learn More
ATCC Corynebacterium glutamicum; 534
Catalog No. 502381761
Encompass
Corynebacterium glutamicum strain 534 [NCIB 10025] is a whole-genome sequenced bacterial type strain. The culture was isolated from sewage and is known to produce glutamic acid. It can be used as a transformation host.
Catalog No. 50-238-1761 Supplier ATCC Supplier No. 13032
Corynebacterium glutamicum strain 534 [NCIB 10025] is a whole-genome sequenced bacterial type strain. The culture was isolated from sewage and is known to produce glutamic acid. It can be used as a transformation host.