Learn More
ATCC Moraxella bovoculi
Catalog No. 502380777
Encompass
Moraxella bovoculi strain 237 is a whole-genome sequenced bacterial type strain that was isolated in 2002 from ocular fluid collected from a calf with infectious bovine keratoconjunctivitis.
Catalog No. 50-238-0777 Supplier ATCC Supplier No. BAA1259
Moraxella bovoculi strain 237 is a whole-genome sequenced bacterial type strain that was isolated in 2002 from ocular fluid collected from a calf with infectious bovine keratoconjunctivitis.